Would you like to log in again?
Ward's® Structure and Function of Mitochondria Lab Activity
Supplier: Avantor
WARD470023-810
470023-810KT 67.99 USD InStock
470023-810
Ward's® Structure and Function of Mitochondria Lab Activity
Educational Classroom Kits and Activities
Observe the “Energy Producers” in Cells
Observe mitochondria both as cytoplasmic organelles and as centers of enzyme activity. Students will prepare microscope slides using celery and a special stain that acts as a hydrogen acceptor; as the dye loses color, students can actually watch the mitochondria under the microscope as they oxidize food. The kit includes two prepared reference slides of plant and animal mitochondria, enough materials for six setups, a teacher’s guide, and student copymaster.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty