Would you like to log in again?
Sundew
Supplier: ROBERT CRESPO
DCAP1
470176-336EA 13.95 USD InStock
470176-336
Sundew
Specimens Live Specimens Plants - Living Material
Drosera sp. Dicot. Rosettes of leaves, radiating from a central stem, trap small insects in leaf hairs, while glands on each tip exude a sticky substance that helps trap the bugs. It is native to acidic bogs in temperate regions and prefers a warm, humid environment. It comes supplied in a pot with care instructions. Size: 2–4".
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty