Would you like to log in again?
Suspension Clamp
Supplier: United Scientific Supplies
CLHK01
470105-132EA 21.95 USD InStock
470105-132
Suspension Clamp
Clamps General Purpose Clamps
Easy to use hook for your support stand to hold small loads.
  • Aluminum bosshead
  • Rod is 10 cm long
Use to suspend a thermometer over a hot plate or for suspending specific heat specimens. Clamp holds items at a distance of 4 1/2" from the support rod. Aluminum hook swivels and locks at any angle.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty