Would you like to log in again?
Ten-Well Polyethylene Test Tube Rack
Supplier: Dynalon
259845
470174-092CS 145 USD InStock
470174-092
Ten-Well Polyethylene Test Tube Rack
Racks Tube Racks
The chemical resistant, lightweight polyethylene racks can withstand temperatures up to 110°C. The wells are rounded and hold tubes up to 18 mm in dia. Size: 9 3/4"L x 2 3/8"W x 3"H.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty