Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Ward's® Termite Symbiosis Kit
Supplier: Avantor
470030-314KT 99.09 USD InStock
470030-314
Ward's® Termite Symbiosis Kit
Educational Classroom Kits and Activities
Investigate Mutualism Between a Protist and an Insect
  • Observe a symbiotic relationship
  • Grades 7-12
  • Includes instructions
  • No reproductive termites are sent (we do not send queens)
Students perform a squash of a termite and prepare a wetmount to observe the protist Trichonympha, which resides in the gut of the termite, where it breaks down cellulose. The kit includes termites and WARD’S Internal Guest of Termites video which demonstrates how to prepare the wetmount. Note: Coupon included for perishable materials. Redeem by phone or e-mail. Some states may require a USDA permit.
Ordering information: No reproductive termites are sent with the culture. We do not send queens.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty