Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tesla Coil Demonstration Apparatus

Supplier: VWR International
PH0841 PH0841AV2
470014-724EA 416.05 USD InStock
470014-724 470221-440
Tesla Coil Demonstration Apparatus
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
The Tesla Coil Demonstration Apparatus transmits electricity without wires.

  • Apparatus Transmits Electricity without Wires
  • Arcs up to 3" Long
  • Potential of 50,000V
  • Safety Guard Included
  • Adjustable Spark Gap Included

This safe source of high-frequency, high-voltage electricity can create arcs up to 3" long.

It has a 50,000V potential.

Ordering information: This kit includes a vibrator, two capacitors, adjustable spark gap, safety guard, primary and secondary, and cord.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty