Would you like to log in again?
Ward's® Testing for Nutrients in Food Lab Activity
Supplier: Avantor
WARD470024-184
470024-184KT 108.99 USD InStock
470024-184
Ward's® Testing for Nutrients in Food Lab Activity
Educational Classroom Kits and Activities
Do You Know What You're Eating?
  • The activity features our exclusive WARD’S®-developed lipid test strips
  • Time requirement: 45 minutes
Perform four qualitative chemical tests on supplied test standards and food samples that you supply in order to determine the presence of macronutrients, such as lipids, sugars, starches, and proteins. Prior to running the experiment students will make predictions on which macronutrients will appear.
Ordering information: Activity includes Glucose Solution, Egg Albumin, Starch Solution,Olive Oil, Biuret Reagent Solution, Benedict's Solution, Iodine Solution, Lipid Test Strips, Disposable Pipettes, Culture Tubes, and Teacher's Guide and Student Copymaster. Hot Plate, Tongs, Timer, Test Tube Racks, Food Sample(s), Marking pen or Wax Pencil, Goggles, Apron, and Protective Gloves are required but not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty