Would you like to log in again?
Vernier® Thermocouple
Supplier: VERNIER SOFTWARE & TECH SE
TCA-BTA
470017-896EA 78.99 USD InStock
470017-896
Vernier® Thermocouple
Temperature Probes
This sensor uses type-K thermocouple wire to measure temperatures over the range of -200 to 1400°C. It can be used to measure flame temperatures as high as 1400°C, or liquid nitrogen temperatures at -196°C. An internal ice-point compensation chip is included, so you do not need to place a reference wire in an ice-water bath. You can simply use one measuring lead to take temperature readings. Each thermocouple is individually calibrated.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty