Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Thermometer Clamp

Supplier: United Scientific Supplies
THCL01
470201-372EA 21.75 USD InStock
470201-372
Thermometer Clamp
Clamps General Purpose Clamps
The easy-adjusting nut on this clamp provides 360° jaw rotation.

  • Maximum Length from Rod to Thermometer: 11 cm (4 ⅖")
  • Clamp Nut Provides 360⁰ Jaw Rotation
  • Easy-Adjusting Nut

The maximum length of this clamp from rod to thermometer is 11 cm (4.4").

The full jaw rotation of this clamp causes prevents hand strain.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty