Would you like to log in again?
Polypropylene Tubing Holders
Supplier: BEL-ART PRODUCTS SE
F18235-0000
470145-012KT 45.85 USD InStock
470145-012
Polypropylene Tubing Holders
Clamps Tubing and Sanitary Clamps
Autoclavable polycarbonate clip that fits over the lip of a beaker, bath, or other vessel.
The 5 cm (2") long arm extends over the beaker and secures the end of a plastic or rubber tube. The maximum size tubing accepted is 9.5 mm ( 3/8") OD.
Packaging: Package of six.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty