Would you like to log in again?
Ultrasonic Cleaner
Supplier: L&R MANUFACTURING COMPANY
311
470105-414EA 1675.9 USD InStock
470105-414
Ultrasonic Cleaner
Ultrasonic Cleaners
Clean the Dirtiest or Most Delicate Laboratory Equipment with Ease.
  • Features include a large tank, on/off switch, and convenient timer
  • Tank features solid-state circuitry
Ultrasonic cleaning systems provide powerful surges of cavitation energy to reach interior areas and the tiniest grooves that hand cleaning can’t touch. Features include a large tank, on/off switch, and convenient timer. Tank is made of durable stainless steel/vinyl-clad metal. Complete with stainless steel lid. Basket not included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty