Would you like to log in again?
Under-Tank Terrarium Heater
Supplier: PET WORLD INC SE
22030
470021-334EA 30.19 USD InStock
470021-334
Under-Tank Terrarium Heater
Animal Supplies Animal Environmental Enrichment
Produces Evenly Distributed Heat Safely
There’s no safer instrument for providing the heat reptiles and other animals require; attached adhesive allows you to simply attach the heater to the underside of the terrarium and plug it in. No wires or other electrical components need to be inserted into the terrarium. Instructions are included. 120 VAC. Size: 8"L x 8 1/4"W. Note: Use only with glass tanks.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty