Would you like to log in again?
Variable Volume Economy Micropipettes
Supplier: United Scientific Supplies
PVV-50 PVV-100 PVV-200 PVV-10 PVV-1000
470123-186EA 236 USD InStock
470123-186 470123-184 470200-654 470123-182 470123-180
Variable Volume Economy Micropipettes
Pipettes Micropipettes
High performance pipette.
  • Compatible with Gilson and eppendorf brand tips
  • Built-in tip ejector
  • Click stop digital system
  • Easy calibration and maintenance
  • Autoclavable tipcone assembly
Easy to disassemble for servicing and autoclaving. The 100 to 1000 μl micropipette uses 1000 μl tips. All other micropipettes use standard 200 μl tips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty