Would you like to log in again?
Velleman DVM810 Multimeter
Supplier: JA ASSOCIATES
DVM 810
470007-276EA 27.7 USD InStock
470007-276
Velleman DVM810 Multimeter
Electrical Meters Digital Multimeters
A compact and easy to use meter for introductory circuits
This economical and compact multimeter hash a 3 1/2 digit LCD for measuring DC and AC voltages, DC currents, resistance, diodes and transistors (hFE), and it features overload protection. The test leads (black and red electrodes) are included.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty