Would you like to log in again?
Ward's® Vermi Composting Kit
Supplier: Avantor
470030-306KT 101.19 USD InStock
470030-306
Ward's® Vermi Composting Kit
Educational Classroom Kits and Activities
Encourage Interest in Recycling with a Tabletop Composter
  • Use worms to break down waste
  • Compare to microbial decomposition
  • Includes a teacher's guide
Investigate the biological breakdown and remediation of human food waste in this stimulating semester-long exercise. Using this process, the degradation will occur much faster than composters based on microbial degradation. Students will use red wiggler worms to efficiently degrade household wastes and create a nutrient-rich soil. The kit includes one composting bin, potting soil, sand, a coupon for red worms, a teacher’s guide, and student copymaster.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty