Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vernier® Blood Pressure Sensor

Supplier: VERNIER SOFTWARE & TECH SE
Vernier®
BPS-BTA
470039-274EA 119.99 USD InStock
470039-274
Vernier® Blood Pressure Sensor
Blood Pressure Monitors
Easily measure blood pressure in the classroom.

  • Measure systolic and diastolic blood pressure, mean arterial pressure, and pulse
  • No stethoscope needed

The blood pressure sensor is a non-invasive sensor designed to measure human blood pressure. It measures systolic, diastolic, and mean arterial pressure utilizing the oscillometric method. Pulse rate is also reported.

Maximum pressure without permanent damage: 1550 mm Hg
Temperature compensated: −20 to +85 °C
Sensing element: SSCMRRN005PGAA5
Combined linearity and hysteresis: typical ±0.25%
Range: 0 mm Hg to 258 mm Hg (pressure range)
Accuracy: ±1 mm Hg
Response time: 1 millisecond

For educational use only.

Accessories information: Small, standard, and large replacement cuffs are available separately.

Ordering information: Includes Vernier® blood pressure sensor, standard adult size adjustable cuff (27 to 39 cm), and bulb pump (with release valve). Required, but not included: LabQuest interface.

Order Now
 

ORDER

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty