Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vernier® Calcium Ion-Selective Electrode

Supplier: VERNIER SOFTWARE & TECH SE
CA-BTA
470120-634EA 298.99 USD InStock
470120-634
Vernier® Calcium Ion-Selective Electrode
Electrodes ISE Electrodes
Measure the concentration of calcium in aqueous samples.

  • Good indicator of hardness of water

Data from the Calcium ISE can give a good indication of hardness of water (as Ca2+). The concentration of Calcium is also used as an endpoint indicator in EDTA-Ca/Mg hardwater titrations. It is important for you to know that Vernier ISE membranes, like all other PVC ISE membranes, have a limited life expectancy; however, the replaceable module of Vernier ISEs allows you to simply discard the used one, and replace it with a new module. We recommend that you do not purchase replacement modules far in advance of their expected time of use, as their life expectancy is limited as well

Note: We recommend that you do not purchase ISE replacement modules too far in advance of their expected time of use as degradation occurs while they are stored on the shelf.

Important: Ion-Selective Electrodes require good chemical technique and careful calibration to obtain accurate results; they are not recommended for middle school or elementary students.

Electrodes are combination-style, non-refillable, and gel-filled
Included with each ISE are two calibration standards and a Short-Term Soaking Bottle
Interfering Ions: Pb2+, Hg2+, Sr2+, Cu2+, Ni2+, NH3, Na+, L1+, K+, Ba2+, Zn2+, Mg2+
Electrode Slope: +26 mV
Electrode Resistance: 100 MΩ
Immersion: 2.8 cm (1.1 in)

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty