Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vernier® Instrumentation Amplifier

Supplier: Vernier
INA-BTA
470120-640EA 88.99 USD InStock
470120-640
Vernier® Instrumentation Amplifier
Power Supplies
The instrumentation amplifier can be used to connect a wide variety of measurement equipment to a Vernier interface and electronically collect, store, and analyze the data.

  • Linearity: 1%
  • Power: 2.5 mA @ 5VDC
  • Frequency response: 0 to 10kHz (f3dB)
  • Impedance : 1MΩ to ground

This newly designed sensor includes auto-ID functionality for each of the six ranges. New input bandwidth of 10 khz allows the capture of higher frequency signals. The Instrumentation Amplifier monitors voltages from 20 mV to 1 V (DC or AC). It has several switch settings to allow you to select the best gain. It is typically used to amplify the chart recorder or analog output of any instrument, such as a third-party gas chromatograph.

It has gain settings of 150, 75, 7.5, 15, 3, and 1.5 ±5% and range setting has 0 to 20 mV, 0 to 200 mV, 0 to 1 V, ±20 mV, ±200 mV, ±1 V.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty