Would you like to log in again?
Vernier® Optics Expansion Kit
Supplier: VERNIER SOFTWARE & TECH SE
OEK
470013-580EA 198.99 USD InStock
470013-580
Vernier® Optics Expansion Kit
Educational Materials Physics Educational Materials Optics Learning Activities
Conduct optics experiments.
  • Light source is LED-based
The parts of the Optics Expansion Kit snap on to the track, so that the parts can be left in place while the track is moved. The three lenses are permanently mounted in three plastic supports. The light source is LED-based, so that it uses less energy and is cool to the touch. A rotating disc on the light source gives you the option of a radiant object, a point source, an “L” with 2 by 1 cm dimension, and parallel lines for resolution experiments. The power supply is the same as the LabQuest power supply. An additional aperture plate is included for experiments based on lens diameter and shape.
For additive and subtractive color mixing experiments, check out the Color Mixer Kit. The Optics Expansion Kit requires a Vernier Track to serve as an optics bench. The Vernier Track is included with a Vernier Dynamics Cart and Track System.
For educational use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty