Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
Vernier® Optics Expansion Kit
Supplier: VERNIER SOFTWARE & TECH SE
PAK-OEK M-OEK
470134-264EA 88.99 USD InStock
470134-264 470133-496
Vernier® Optics Expansion Kit
Educational Materials Physics Educational Materials Optics Learning Activities
Add the Vernier Optics Expansion Kit to your Vernier Dynamics System to conduct optics experiments, such as image formation with lenses and light intensity vs. distance. You can even use the kit to build a basic telescope.
The Optics Expansion Kit includes:
-Combination LED point and luminous light source with power supply
-Three lenses integrated with holders (100, 200 and —150 mm focal length)
-Screen and holder
-Light Sensor holder
-Aperture screen and holder
-Power supply
The parts of the optics expansion kit snap on to the track, so that the parts can be left in place while the track is moved. The three lenses are permanently mounted in three plastic supports.
The light source is LED-based, so that it uses less energy and is cool to the touch. A rotating disc on the light source gives you the option of a radiant object, a point source, an 'L' with 2 by 1 cm dimension, and parallel lines for resolution experiments. The power supply is the same as the LabQuest power supply. An additional aperture plate is included for experiments based on lens diameter and shape.
Extend your optics lessons to allow students to study polarization of light. Set includes a fixed polarizer, an adjustable analyzer, and an adjustable analyzer for rotary motion sensor. Create a complete system for studying polarization with the optics expansion kit, plus a rotary motion sensor, and light sensor, each available separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty