Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vernier® Photogate

Supplier: Vernier
VPG-BTD
470002-966EA 54.99 USD InStock
470002-966
Vernier® Photogate
Educational Materials Physics Educational Materials Mechanics Learning Activities
Study free fall, rolling objects, collisions, and pendulums.

  • Includes an accessory rod for mounting to a ring stand

Photogates allow for extremely accurate timing of events within physics experiments, for studying free fall, air track collisions, pendulum periods, the speed of a rolling object, among other things.

The gate has an input port so multiple gates can be connected in a daisy-chain configuration with up to four gates going to a single interface channel.

The laser gate mode requires the addition of a common pen laser, which is directed into the laser port. The laser may be some distance from the gate, so that you can measure the speed of larger objects such as a rocket, model car, or even a real automobile.

Power req: 5 VDC at 40 mA
Infrared source: Peak at 880 nm
Output is high and LED off for unblocked gate
Output is low and LED on for blocked gate
Beam approximately 9 mm from the end of the gate arms

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty