Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Vernier® Picket Fence

Supplier: Vernier
PF
470003-068EA 9.99 USD InStock
470003-068
Vernier® Picket Fence
Educational Materials Physics Educational Materials Mechanics Learning Activities
Free fall experiments have never been easier!

  • Each opaque bar is 25 mm wide, with 25 mm between opaque bars

The Vernier picket fence has eight opaque bars spaced every 5 cm, silk-screened directly onto clear plastic. Drop the picket fence through a photogate to obtain records of position, velocity and acceleration vs time or to measure g.

Ordering information: Vernier photogates available separately.

Packaging: Shipped with protective backing. Remove protective backing (may be on both sides) before use.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty