Would you like to log in again?
Vernier® Respiration Monitor Belt
Supplier: VERNIER SOFTWARE & TECH SE
RMB
470001-866EA 62.99 USD InStock
470001-866
Vernier® Respiration Monitor Belt
Pressure Sensors
The respiration monitor belt is used with our gas pressure sensor to measure human respiration.
  • Respiration rate calculation is only supported in Logger Pro and LabQuest App.
Simply strap the belt around your chest, then pump air into the belt with the hand bulb. You can then monitor the pressure associated with the expansion and contraction of your chest during breathing. (Requires the Gas Pressure Sensor.)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty