Would you like to log in again?
Washington School Collection
Supplier: Avantor
WARD470025-176 WARD470025-178 WARD470025-174
470025-174EA 109.49 USD InStock
470025-174 470025-176 470025-178
Washington School Collection
Specimens Rock Specimens Rock Collections
A Comprehensive Classroom Survey Set of Minerals and Rocks
  • Descriptions of each specimen
  • Hand cut
  • Housed in a compartmented box
An exceptional value for a quality collection. Originally developed by Ward's®, over the years the Washington School Collection has become one of the most widely accepted secondary-school earth science teaching aids. Carefully selected, high-quality rock and mineral samples provide students with an excellent introduction to earth materials and their characteristics. The set is housed in a compartmented collection box, and descriptions of each specimen are printed on a label in the lid of the box.
Contents:
40 numbered samples, 1×2".
Ordering information: Mineral contents include Rock-Forming (calcite, garnet, hornblende, microcline, milky quartz, muscovite, plagioclase feldspar, quartz [rock crystal]), Metallic Ore (bauxite, chalcopyrite, galena, hematite, limonite, magnetite, sphalerite), and Industrial (graphite, gypsum, halite, pyrite, talc). Rock contents include Igneous (basalt, gabbro, granite, obsidian, rhyolite, syenite), Sedimentary (argillaceous sandstone, bituminous coal, chert, conglomerate, dolomitic limestone, gravel, red sandstone, shale, shell limestone), and Metamorphic (gneiss, marble, quartzite, schist, slate).
Caution: Due to the nature of this product it may contain asbestos.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty