Would you like to log in again?
Ward's® What's Up With the Ozone Lab Activity
Supplier: Avantor
WARD470175-696
470175-696EA 133.99 USD InStock
470175-696
Ward's® What's Up With the Ozone Lab Activity
Educational Classroom Kits and Activities
Test Your Local Area for Ozone Conditions
  • Designed for AP level students
  • Practice preparing test strips
The differentiation of ozone levels within the same general vicinity is explored in this AP level lab. Students prepare their own test strips and carry out tests for tropospheric ozone pollution. They then analyze the results for local variation and possible impact on human health. Includes a teacher’s guide, student copymasters, and materials for six setups.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty