Would you like to log in again?
Ward's® Why Cells Shrink and Swell Lab Activity
Supplier: Avantor
470024-186KT 76.69 USD InStock
470024-186
Ward's® Why Cells Shrink and Swell Lab Activity
Educational Classroom Kits and Activities
Large Plant and Animal Cells Provide a Great View of Osmosis in Action
Students will observe the effects of osmotic imbalance in just minutes, and in the process learn how and why cells passively maintain homeostasis. They can view under a microscope (available separately) both turgor and plasmolysis in plant cells and hemolysis and crenation in bovine red blood cells. Also, an interactive activity guides students as they transform your classroom into a living cell to further explore the effects of osmotic imbalance. It includes enough materials for 15 setups, a teacher’s guide, and student copymaster. Note: Coupon included for perishable materials. Redeem by mail, fax, phone, or e-mail.
Materials Included
•1 Package, microscope slides
•1 Package, coverslips
•3 Culture tubes with cap
•1 Package, sodium chloride (for a 5% solution)
•1 Bottle, physiological saline (0.9%), 500 ml
•30 Disposable pipets
•1 Biohazard bag
•1 Redemption coupon for: 15 Elodea sprigs, 10 ml Vial, bovine blood
Materials Needed But Not Provided
•Test tube racks
•1 L beaker
•Paper towels
•Microscope(s)
•Distilled water
•Forceps
•Marking pen or wax pencil
•Apron
•Gloves
•Goggles
Time Requirement
•45 Minutes
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty