Would you like to log in again?
Wide Mouth, Safety Labeled Washing Bottles
Supplier: BEL-ART PRODUCTS SE
F11646-0638
470123-964PK 51.3 USD InStock
470123-964
Wide Mouth, Safety Labeled Washing Bottles
Bottles Wash Bottles
Polyethylene bottles with highly visible, permanently silk-screened labels provide easyidentification of common laboratory solutions. Diamond Alert symbols indicate thepotential hazards of the contents. Bottle capacity: 500 ml. With color-coded polyallomernozzle/caps.
Packaging: Package of six.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty