Would you like to log in again?
Wind Powered Generator
Supplier: THE SCIENCE SOURCE SE
800
470008-040EA 536 USD InStock
470008-040
Wind Powered Generator
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
A superior teaching aid, this generator incorporates new opportunities for student experiments in aerodynamic electricity transfer. The generator blade assembly consists of six vanes, 37cm in diameter, with a total sail area of 77cm2. This is connected to a DC generator and 1.5 volt bulb to indicate electrical output. Other features include a full cover for outdoor use, a weighted base, a structural aluminum tower, and a wind-seeking, balanced tail assembly. The unit may be used outside, or inside with an air supply. Comes complete with instruction sheets and teacher’s manual. Size: 48cm H.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty