Would you like to log in again?
Ward's® World Beneath Our Feet Lab Activity
Supplier: Avantor
WARD470023-828
470023-828KT 169.99 USD InStock
470023-828
Ward's® World Beneath Our Feet Lab Activity
Educational Classroom Kits and Activities
Look for Macro- and Microorganisms in Soil Samples Your Class Collects
  • Observe soil microorganisms
  • Practice sterile lab techniques
  • Includes a teacher's guide
Under our feet is a whole world of organisms your students might never see. You can show your students this hidden world with our lab activity. After sifting through soil for macroscopic creatures, students use Gram staining techniques and examine the soil for microscopic creatures. The kit comes with enough materials for ten setups, a teacher’s guide, and student copymaster.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty