Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Estes Yankee Model Rocket

Supplier: Avantor
Estes
1381
470006-852CS 426.1 USD InStock
470006-852
Estes Yankee Model Rocket
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Easy-to-assemble intermediate-level rocket with laser cut balsa fins.

  • Intermediate-level build
  • Requires standard size 18 mm A, B, or C engines
  • Max. altitude 600 m (2000')

This rocket is easy to build, yet it delivers an impressive performance. Its 11" body makes it lightweight. The 5" streamer landing system helps ensure this high-flying rocket lands safely without drifting too far.

Ordering information: Launch system and rocket engines available separately.
Single kit includes all parts needed to assemble (1) model rocket. Some tools and glue may be required.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty