Would you like to log in again?
BluePhos® Microwell Substrate Kit, KPL
Supplier: KPL
5120-0061 5120-0059 5120-0060
95059-224EA 155.46 USD InStock
95059-224 95059-222 95059-220
BluePhos® Microwell Substrate Kit, KPL
Assays ELISAs
Excellent alternative to pNPP
BluePhos substrate provides greater sensitivity than traditional pNPP substrates as well as excellent linearity. Upon stopping the substrate converts from yellow to purple.
Designed for use with AP conjugates. A soluble, proprietary form of BCIP (5-bromo-4-chloro-3-indoyl phosphate) that features greater sensitivity, wider working range, lower background, and more convenience than pNPP.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty