Would you like to log in again?
Protein Detector™ Peroxidase ELISA Kits, Clinical Diaganostics, Inc
Supplier: LGC Clinical Diagnostics
Protein Detector™
5110-0010 5110-0011 5110-0009
95059-322EA 519.33 USD InStock
95059-322 95059-326 95059-324
Protein Detector™ Peroxidase ELISA Kits, Clinical Diaganostics, Inc
Assays ELISAs
HRP ELISA Kit offer a convenient starting point for the development of microwell ELISA protocols for antigen and antibody detection
Kit contains Coating Solution Concentrate; BSA Diluent/Blocking Solution Concentrate; Wash Solution Concentrate; 50% Glycerol Solution; HRP labeled Goat Anti-Mouse IgG (H+L); ABTS Peroxidase Substrate System; and ABTS Stop Solution.
Highly sensitive reagents are optimized for antigen detection in picogram quantities. The wash and block solutions ensure low background. This kit can be used for direct and indirect detection.
The kit provides pretested components and suggested protocols to facilitate the development of ELISA procedures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty