Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
HyClone™ AdvanceSTEM Human Somatic Stem Cell Media, HyClone products (Cytiva)
Supplier: HyClone products (Cytiva)
HyClone™
SH30889.02 SH30886.02 SH30881.02 SH30886.02
95053-246EA 765.2 USD InStock
CA95053-244 95053-246 95053-244 95053-242
HyClone™ AdvanceSTEM Human Somatic Stem Cell Media, HyClone products (Cytiva)
Cell Culture Media
HyClone AdvanceSTEM Human Somatic Stem Cell Media has been developed for mesenchymal stem cell research. Support applications ranging from the expansion and maintenance of human adult mesenchymal stem cells, to the directed differentiation of these cells into such cell types as neurons, adipocytes, chondrocytes, and osteocytes.
Adipogenic Differentiation Medium supports adipogenic differentiation from human adipose-derived and bone marrow-derived mesenchymal stem cells. Chondrogenic Differentiation Medium supports differentiation of a variety of hMSCs into chondrocytes. Osteogenic Differentiation Medium supports the differentiation of a variety of human mesenchymal stem cells, including human adipose-derived, Multipotent Cord.
Support applications ranging from the expansion and maintenance of human adult mesenchymal stem cells, to the directed differentiation of these cells into such cell types as neurons, adipocytes, chondrocytes, and osteocytes.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty