Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1H,1H,9H,9H-Perfluoro-1,9-nonanediol ≥90%

Supplier: Matrix Scientific
Warning
Synonyms: 1H,1H,9H,9H-Perfluoro-1,9-nonanediol2,2,3,3,4,4,5,5,6,6,7,7,8,8-Tetradecafluoro-1,9-nonanediol2,2,3,3,4,4,5,5,6,6,7,7,8,8-Tetradecafluoro-1,9-nonandiol

001216-5G 001216-25G
101781-180EA 186.15 USD InStock
101781-180 101781-178
1H,1H,9H,9H-Perfluoro-1,9-nonanediol ≥90%
1H,1H,9H,9H-Tetradecafluoro-1,9-nonandiol


Formula: C₉H₆F₁₄O₂
MW: 412.12 g/mol
Melting Pt: 108-112 °C
MDL Number: MFCD00153275
CAS Number: 203303-01-9
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty