Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

trans-2,6-Difluorocinnamic acid ≥98%

Supplier: Matrix Scientific
Warning
Synonyms: (E)-3-(2,6-Difluorophenyl)acrylic acid

001528-100G 001528-5G 001528-25G
101781-900EA 1361.54 USD InStock
101781-900 101781-898 101781-896
trans-2,6-Difluorocinnamic acid ≥98%
trans-2,6-Difluorocinnamic acid
Hazard code(s): Irritant


Formula: C₉H₆F₂O₂
MW: 184.14 g/mol
Melting Pt: 124…125 °C
MDL Number: MFCD00010319
CAS Number: 102082-89-3
Order Now
Specification Test Results
Melting point 124-125°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty