Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-4-fluoroanisole ≥97%

Supplier: Matrix Scientific
Warning
Synonyms: 2-Bromo-4-fluoro-1-methoxybenzene

001689-100G 001689-25G
101782-282EA 36.92 USD InStock
101782-282 101782-284
2-Bromo-4-fluoroanisole ≥97%
2-Bromo-4-fluoroanisole
Hazard code(s): Irritant


Formula: C₇H₆BrFO
MW: 205.03 g/mol
Boiling Pt: 89 °C (1mm)
Melting Pt: –27 °C
Density: 1.581 g/cm³
MDL Number: MFCD00012014
CAS Number: 452-08-4
Order Now
Specification Test Results
Boiling point 89°C/1mm
Density 1.581
ND20 1.545

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty