Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Chloro-3-fluorophenol ≥98%

Supplier: Matrix Scientific

002350-25G 002350-5G
101783-718EA 67.69 USD InStock
101783-718 101783-716
4-Chloro-3-fluorophenol ≥98%
4-Chloro-3-fluorophenol
Hazard code(s): Corrosive


Formula: C₆H₄ClFO
MW: 146.55 g/mol
Boiling Pt: 84 °C (44mm)
Melting Pt: 54…56 °C
MDL Number: MFCD00042583
CAS Number: 348-60-7
UN: 3261
ADR: 8,III
Order Now
Specification Test Results
Boiling point 84°C/44mm
Melting point 54-56°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty