Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Fluorobenzyl alcohol ≥98%

Supplier: Matrix Scientific
Synonyms: (3-Fluorophenyl)methanol

003322-100G 003322-25G
101785-570EA 32.31 USD InStock
101785-570 101785-572
3-Fluorobenzyl alcohol ≥98%
3-Fluorobenzyl alcohol
Hazard code(s): Irritant


Formula: C₇H₇FO
MW: 126.13 g/mol
Boiling Pt: 104…105 °C (22mm)
Density: 1.164 g/cm³
MDL Number: MFCD00004631
CAS Number: 456-47-3
EINECS: 207-265-3
Order Now
Specification Test Results
Boiling point 104-105°C/2
Density 1.164
ND20 1.51

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty