Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluorobenzyl alcohol ≥98%

Supplier: Matrix Scientific
Synonyms: 2-Fluorobenzyl Alcohol(2-Fluorophenyl)methanol

003323-25G 003323-100G
101785-574EA 30.77 USD InStock
101785-574 101785-576
2-Fluorobenzyl alcohol ≥98%
2-Fluorobenzyl alcohol
Hazard code(s): Irritant


Formula: C₇H₇FO
MW: 126.13 g/mol
Boiling Pt: 199…200 °C
Density: 1.173 g/cm³
MDL Number: MFCD00004601
CAS Number: 446-51-5
EINECS: 207-170-7
Order Now
Specification Test Results
Boiling point 199-200°C
Density 1.173
ND20 1.514

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty