Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(2-Fluorophenyl)ethanol ≥99%

Supplier: Matrix Scientific
Synonyms: 2-Fluorophenethyl alcohol

003851-5G 003851-25G
101786-308EA 87.69 USD InStock
101786-308 101786-306
2-(2-Fluorophenyl)ethanol ≥99%
2-(2-Fluorophenyl)ethanol
Hazard code(s): Irritant


Formula: C₈H₉FO
MW: 140.16 g/mol
Boiling Pt: 104…106 °C (14 mmHg)
Density: 1.045 g/cm³
MDL Number: MFCD00002887
CAS Number: 50919-06-7
Order Now
Specification Test Results
Density 1.045
ND20 1.509

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty