Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,4-Dibromo-2,5-difluorobenzene ≥98%

Supplier: Matrix Scientific
Warning

004536-100G 004536-25G
101787-424EA 150.77 USD InStock
101787-424 101787-422
1,4-Dibromo-2,5-difluorobenzene ≥98%
1,4-Dibromo-2,5-difluorobenzene
Hazard code(s): Irritant


Formula: C₆H₂Br₂F₂
MW: 271.89 g/mol
Boiling Pt: 96 °C (20mm)
Melting Pt: 60…62 °C
MDL Number: MFCD00000346
CAS Number: 327-51-5
EINECS: 206-317-2
Order Now
Specification Test Results
Boiling point 96°C/20mm
Melting point 60-62°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty