Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Bromobenzyl alcohol ≥98%

Supplier: Matrix Scientific
Synonyms: (3-Bromophenyl)methanol

004755-100G
101787-780EA 106.15 USD InStock
101787-780
3-Bromobenzyl alcohol ≥98%
3-Bromobenzyl alcohol
Hazard code(s): Irritant


Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 165 °C (16mm)
Density: 1.560 g/cm³
MDL Number: MFCD00004629
CAS Number: 15852-73-0
EINECS: 239-975-4
Order Now
Specification Test Results
Boiling point 165°C/16mm
Density 1.56
ND20 1.584

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty