Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

α,α,α-Trifluoro-3-tolualdehyde ≥97%

Supplier: Matrix Scientific
Warning
Synonyms: Cinacalcet Impurity 233-(Trifluoromethyl)benzaldehyde

005504-100G 005504-25G
101789-074EA 83.08 USD InStock
101789-074 101789-072
α,α,α-Trifluoro-3-tolualdehyde ≥97%
α,α,α-Trifluoro-3-tolualdehyde
Hazard code(s): Irritant


Formula: C₈H₅F₃O
MW: 174.12 g/mol
Boiling Pt: 83…86 °C (30mm)
Density: 1.301 g/cm³
MDL Number: MFCD00003373
CAS Number: 454-89-7
EINECS: 207-228-1
Order Now
Specification Test Results
Boiling point 83-86°C/30m
Density 1.301
ND20 1.465

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty