Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4-Dichloropyrimidine ≥98%

Supplier: Matrix Scientific
Warning

005611-100G
101789-236EA 75.38 USD InStock
101789-236
2,4-Dichloropyrimidine ≥98%
2,4-Dichloropyrimidine
Hazard code(s): Irritant, Refrigerate


Formula: C₄H₂Cl₂N₂
MW: 148.98 g/mol
Boiling Pt: 101 °C (23mm)
Melting Pt: 58…60 °C
MDL Number: MFCD00006061
CAS Number: 3934-20-1
EINECS: 223-508-6
Order Now
Specification Test Results
Boiling point 101°C/23mm
Melting point 58-60°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty