Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl propionylacetate ≥96%

Supplier: Matrix Scientific
Synonyms: Ethyl 3-oxopentanoateEthyl 3-oxovalerate

005735-25G
101789-464EA 84.62 USD InStock
101789-464
Ethyl propionylacetate ≥96%
Ethyl propionylacetate
Hazard code(s): Irritant


Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Boiling Pt: 83…84 °C (12mm)
Density: 1.01 g/cm³
MDL Number: MFCD00009317
CAS Number: 4949-44-4
EINECS: 225-593-5
Order Now
Specification Test Results
Boiling point 83-84°C/12m
Density 1.01
ND20 1.422

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty