Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,6-Bis(trifluoromethyl)benzoic acid ≥98%

Supplier: Matrix Scientific

005806-5G 005806-1G
101789-556EA 136.92 USD InStock
101789-556 101789-554
2,6-Bis(trifluoromethyl)benzoic acid ≥98%
2,6-Bis(trifluoromethyl)benzoic acid
Hazard code(s): Irritant


Formula: C₉H₄F₆O₂
MW: 258.12 g/mol
Boiling Pt: 260 °C (1013 hPa)
Melting Pt: 138…140 °C
MDL Number: MFCD00000376
CAS Number: 24821-22-5
EINECS: 246-479-1
Order Now
Specification Test Results
Melting point 138-140°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty