Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Bis(trifluoromethyl)phenol ≥95%

Supplier: Matrix Scientific
Synonyms: α,α,α,α',α',α'-Hexafluoro-3,5-xylenol

005884-25G
101789-694EA 58.46 USD InStock
101789-694
3,5-Bis(trifluoromethyl)phenol ≥95%
3,5-Bis(trifluoromethyl)phenol
Hazard code(s): Irritant


Formula: C₈H₄F₆O
MW: 230.11 g/mol
Boiling Pt: 97 °C (50mm)
Melting Pt: 20…21 °C
Density: 1.511 g/cm³
MDL Number: MFCD00000386
CAS Number: 349-58-6
EINECS: 206-488-3
Order Now
Specification Test Results
Boiling point 97°C/50mm
Melting point 20-21°C
Density 1.511
ND20 1.415

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty