Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tin (II) trifluoromethanesulfonate ≥90%

Supplier: Matrix Scientific
Synonyms: Tin trifluoromethanesulfonateTin(II) trifluoromethanesulfonateTin triflateTin(II) trifluoromethanesulphonateTin trifluoromethanesulphonate

006376-5G 006376-25G 006376-1G
101790-452EA 155.38 USD InStock
101790-452 101790-454 101790-450
Tin (II) trifluoromethanesulfonate ≥90%
Tin (II) trifluoromethanesulphonate
Hazard code(s): Toxic, Moisture Sensitive


Formula: C₂F₆O₆S₂Sn
MW: 416.85 g/mol
MDL Number: MFCD00191251
CAS Number: 62086-04-8
UN: 3288
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty