Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(Methylthio)phenol ≥98%

Supplier: Matrix Scientific
Warning
Synonyms: 2-Hydroxythioanisole

006574-5G 006574-100G 006574-25G
101790-782EA 29.23 USD InStock
101790-782 101790-784 101790-786
2-(Methylthio)phenol ≥98%
2-(Methylthio)phenol
Hazard code(s): Irritant, Stench


Formula: C₇H₈OS
MW: 140.21 g/mol
Boiling Pt: 104…106 °C (22mm)
Density: 1.168 g/cm³ (25 °C)
MDL Number: MFCD00002211
CAS Number: 1073-29-6
EINECS: 214-027-2
Order Now
Specification Test Results
Boiling point 104-106°C/2

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty