Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-4-methylpyridine ≥97%

Supplier: Matrix Scientific
Warning
Synonyms: 2-Chloro-4-picoline

007067-25G 007067-100G
101791-954EA 87.69 USD InStock
101791-954 101791-952
2-Chloro-4-methylpyridine ≥97%
2-Chloro-4-methylpyridine
Hazard code(s): Irritant, Irritant-Harmful


Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 40 °C (0.05mm)
Melting Pt: 115 °C
Density: 1.456 g/cm³ (25 °C)
MDL Number: MFCD00023418
CAS Number: 3678-62-4
EINECS: 222-951-2
Order Now
Specification Test Results
Boiling point 40°C/0.05mm

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty